Learn More
Medchemexpress LLC 2-chloro-N-(3,4-dimethoxybenzyl)acetamide | 65836-72-8 | MFCD01733639 | 98.8% | 243.69 g·mol⁻1 | C11H14ClNO3 | 500 MG

Supplier: Medchemexpress LLC HY101836500MG
DKM 2-93 is a covalent small-molecule inhibitor of ubiquitin-like modifier activating enzyme 5 (UBA5). It inhibits UBA5 by covalently modifying the catalytic cysteine and has shown biochemical and cellular activity in preclinical pancreatic cancer models.
- Covalent UBA5 inhibitor (IC50 430 μM).
- Demonstrated cellular activity (PaCa2 EC50 90 μM; Panc1 EC50 30 μM).
- High reported purity (98.8%).
- Chemical formula C11H14ClNO3; molecular weight 243.69 g·mol⁻1.
- Solid, white to light yellow appearance; available in multiple package sizes including 500 MG.
- Recommended storage: powder -20°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.