Learn More
Medchemexpress LLC 2-chloroethyl ethylsulfide | 693-07-2 | MFCD00000979 | 97.3% | 124.63 | C4H9ClS | 5 G
SDP
Supplier:  Medchemexpress LLC HYW1991905G
Encompass_Preferred
2-CEES is a sulfur mustard analog used as a research reagent to model alkylation-induced DNA damage. It selectively forms DNA monoadducts and has been reported to induce centrosome amplification and chromosome instability in mammalian cells. Intended for laboratory research use only; handle under appropriate safety protocols and store under recommended conditions.
  • Forms DNA monoadducts relevant to alkylation studies.
  • Useful for investigating centrosome amplification and chromosomal instability.
  • Available as a liquid in multiple package sizes for lab-scale experiments.
  • High purity suitable for research applications.
  • Requires controlled storage and handling under inert atmosphere.
Catalog No. 50-002-80778