Learn More
Medchemexpress LLC 2-Chloronicotinaldehyde | 36404-88-3 | 99.76% | 141.56 | 50 G
Supplier: Medchemexpress LLC HY3279150G
2-Chloropyridine-3-carbaldehyde is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
- Biochemical reagent for life science related research
- Used as a biological material or organic compound
- Exhibits cytotoxicity against mouse B16F10 cells