Learn More
Medchemexpress LLC 2-chloronicotinic acid | 2942-59-8 | MFCD00006236 | 99.8% | 157.56 g/mol | C6H4ClNO2 | 500 G

Supplier: Medchemexpress LLC HYW017249500G
2-Chloronicotinic acid (CAS 2942-59-8) is a chlorinated pyridine-carboxylic acid used as an intermediate and biochemical reagent in organic synthesis and life-science research. It serves as a versatile building block for heterocycle construction and assay development.
- High purity suitable for synthesis and analytical applications.
- Molecular weight 157.56 g/mol.
- Chemical formula C6H4ClNO2.
- Versatile building block for medicinal chemistry and heterocycle synthesis.
- Available in multiple pack sizes to support small and large scale work.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.