Learn More
Medchemexpress LLC 2-chlorophenothiazine | 92-39-7 | MFCD00005016 | 50mg
SDP
Supplier:  Medchemexpress LLC HYY174950MG
Encompass_Preferred
2-Chlorophenothiazine is a phenothiazine derivative 2-Chlorophenothiazine has the activity of inhibiting DNA RNA and protein synthesis 2-Chlorophenothiazine is an intermediate in the synthesis of Chlorpromazine (HY-12708) and Perphenazine (HY-A0077)[1]
Catalog No. 50-002-66369