Learn More
Medchemexpress LLC (2-chlorophenyl)(phenyl)methanone | 5162-03-8 | MFCD00000558 | 98.8% | 216.66 g/mol | C13H9ClO | 50 G

Supplier: Medchemexpress LLC HYW01411450G
(2-Chlorophenyl)(phenyl)methanone is a white to off-white solid used as an organic reagent and intermediate for life-science and synthetic chemistry research. It is supplied at high purity for laboratory use and intended for research applications only.
- White to off-white solid physical form.
- High purity: 98.8%.
- Molecular formula C13H9ClO; molecular weight 216.66 g/mol.
- Suitable as a building block and reagent in organic synthesis and biochemical studies.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Available in multiple pack sizes including 10 g, 25 g, 50 g, 100 g, 500 g, and 1 kg.
- For research use only; not for human or diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.