Learn More
Medchemexpress LLC 2-Chlorophenylglycine | 88744-36-9 | 185.61 | 500 G

Supplier: Medchemexpress LLC HYW002235500G
2-Chlorophenylglycine is an amino acid derivative of Glycine, supplied as a white to off-white solid with 99.82% purity. This compound has potential as an ergogenic dietary substance, influencing anabolic hormone secretion, supporting fuel supply during exercise, enhancing mental performance under stress, and helping prevent exercise-induced muscle damage. For research use only.
- Chemical properties:
- Molecular weight: 185.61
- Formula: C8H8ClNO2
- CAS no.: 88744-36-9
- Appearance: Solid, white to off-white
- Purity: 99.82%
- Storage and handling:
- Shipping: Room temperature (continental US)
- Powder: -20°C for 3 years, 4°C for 2 years
- In solvent: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.