Learn More
Medchemexpress LLC 2-Chloropyrimidine-5-carboxylic acid | 374068-01-6 | 100.0% | C₅H₃ClN₂O₂ | 500 G

Supplier: Medchemexpress LLC HY69142500G
2-Chloropyrimidine-5-carboxylic acid is a drug intermediate used for the synthesis of various active compounds.
- Solid appearance
- White to light yellow color
- Soluble in DMSO at 100 mg/mL
- Recommended storage at 4°C under nitrogen
- Recommended storage in solvent at -80°C for 6 months or -20°C for 1 month under nitrogen
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.