Learn More
Medchemexpress LLC 2-Chloropyrimidine-5-carboxylic acid | 374068-01-6 | 99.9% | C5H3ClN2O2 | 50 G

Supplier: Medchemexpress LLC HY6914250G
2-Chloropyrimidine-5-carboxylic acid is a chemical compound primarily used as a drug intermediate for the synthesis of various active compounds. It typically appears as a white to light yellow solid.
- Intermediate for active compound synthesis
- White to light yellow solid
- Molecular weight: 158.54
- Soluble in DMSO (100 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.