Learn More
Apexbio Technology LLC 2-Cl-IB-MECA 163042-96-4 10mg
SDP
Supplier:  Apexbio Technology LLC B659710
Encompass_Preferred
2-Cl-IB-MECA is a small-molecule agonist targeting the adenosine A3 receptor (A3AR) It is designed to selectively activate A3AR thereby regulating cellular processes such as inflammation modulation and cytoprotection 2-Cl-IB-MECA exerts its biological activity primarily through activation of A3AR-mediated signaling pathways Based on these pharmacological properties 2-Cl-IB-MECA holds research potential in chronic inflammatory responses tissue remodeling hepatic fibrosis oncology and inflammatory disorder contexts and is commonly used to evaluate receptor-mediated biochemical pathways in in vitro cellular models and experimental animal studies
Catalog No. 50-000-92103