Learn More
Medchemexpress LLC 2-CTC Resin | 934816-82-7 | 50 G
SDP
Supplier:  Medchemexpress LLC HY14210450G
Encompass_Preferred
2-CTC Resin is a solid-phase synthesis carrier used in studies of metabolic disorders in prostate cancer cells. It functions as a reaction solution for terminal residues and adenine nucleotides to form cyclic peptides. This resin prevents racemization during the incorporation of the first protected amino acid and minimizes diketopiperazine formation, making it a versatile resin for large-scale peptide production.
  • Utilized for studying metabolic disorders in prostate cancer cells
  • Functions as a reaction solution for terminal residues and adenine nucleotides
  • Forms cyclic peptides
  • Prevents racemization during incorporation of the first protected amino acid
  • Minimizes diketopiperazine formation
  • One of the most widely used and versatile resins for large-scale peptide production
  • Mesh size: 100-200 mesh
  • DVB percentage: 1% DVB
  • Loading capacity: 0.4-3.0 mmol/g
Catalog No. 50-003-53689