Learn More
Medchemexpress LLC 2-Cyanothiazole | 1452-16-0 | 110.14 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW003625
Encompass_Preferred
2-Cyanothiazole is a drug intermediate for the synthesis of various active compounds. It is for research use only and not sold to patients. It appears as a solid with a color ranging from light yellow to light brown.
  • Drug intermediate for synthesis of various active compounds
  • For research use only
  • Appears as a solid
  • Color ranges from light yellow to light brown
Catalog No. 50-004-48215