Learn More
Medchemexpress LLC 2-Cyclopropylacetic acid | 5239-82-7 | 99.96% | 100.12 | 500 G

Supplier: Medchemexpress LLC HYW010610500G
2-Cyclopropylacetic acid is a drug intermediate used in the synthesis of various active compounds. It appears as a colorless to light yellow liquid. This product is intended for research use only and is not sold to patients. The density is 1.172±0.06 g/cm3 and its molecular formula is C5H8O2.
- Suitable for high-speed applications
- Colorless to light yellow liquid appearance
- Density of 1.172±0.06 g/cm3
- Molecular formula C5H8O2
- For research use only
- Pure form stable at -20°C for 3 years or 4°C for 2 years
- In solvent, stable at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.