Learn More
Cayman Chemical 2-deoxy-2-fluoro L-Fucose 5mg
SDP
Supplier:  Cayman Chemical 171715 MG
Encompass_Preferred
A fluorinated fucose analog that can be metabolized inside the cell to a substrate-based inhibitor of fucosyltransferases
Catalog No. 50-296-5969