Learn More
Selleck Chemical LLC 2-Deoxy-5-Fluorocytidine S2477-5mg
SDP
Supplier:  Selleck Chemical LLC S24775MG
Encompass_Preferred
2 -Deoxy-5-fluorocytidine (5-fluoro-2( )-deoxycytidine FCdR) a pyrimidine analog is a DNA methyltransferase (DMNT) inhibitor currently in clinical trials for solid tumors
Catalog No. 50-311-2147