Learn More
Selleck Chemical LLC 2-Deoxy-D-glucose (2-DG) 100mg 154-17-6 2-deoxyglucose, NSC 15193
SDP
Supplier:  Selleck Chemical LLC S4701500MG
Encompass_Preferred
2-Deoxy-D-glucose (2-DG, 2-deoxyglucose, NSC 15193), an analog of glucose, is a glycolytic inhibitor with antiviral activity. 2-Deoxy-D-glucose induces apoptosis and inhibits Herpes Simplex Virus type-1 (HSV-1) receptor expression. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4198