Learn More
Abcam 2'-Deo x yadenosine 5'-monophosphate (dAMP) free acid, nucleotide derivative, 250MG
Supplier:  Abcam AB146524250MG
Encompass_Preferred
MW 331.2 g/mol, Purity >98%. Nucleotide derivative, used to study adenosine-based interactions during DNA synthesis, inhibition, and damage.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4597