Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-deoxyadenosine | 958-09-8 | MFCD00005754 | 99.9% | 251.24 | C10H13N5O3 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYW040329R10MG

Encompass_Preferred

2'-deoxyadenosine is an analytical standard of the nucleoside 2'-deoxyadenosine supplied for research and analytical applications, providing a high-purity reference material for assay calibration and method development.

  • High purity: 99.9%.
  • Cas number: 958-09-8.
  • Molecular formula: C10H13N5O3.
  • Molecular weight: 251.24.
  • Available in small pack sizes suitable for analytical use, e.g., 10 MG.
  • Intended for assay calibration, quality control, and analytical method development.

Catalog No. 50-002-09339


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.