Learn More
Medchemexpress LLC 2'-Deoxycytidine-13C9,15N3 | 1987883-22-6 | 99.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYD0184S31MG
Encompass_Preferred
2'-Deoxycytidine-13C9,15N3 is a 13C and 15N-labeled 2'-Deoxycytidine. This deoxyribonucleoside can inhibit the biological effects of Bromodeoxyuridine (Brdu) and is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-10267