Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'-Deoxyguanosine monohydrate | 312693-72-4 | 99.7% | 1 G
SDP

Supplier:  Medchemexpress LLC HYD01851G

Encompass_Preferred

2'-Deoxyguanosine monohydrate is a nucleoside composed of guanine and 2'-deoxyribose, serving as one of the basic structural units of DNA. Its oxidation products may participate in DNA damage, which can affect gene expression or induce cancer.

  • Purity: 99.70%
  • Appearance: Solid
  • Color: White to off-white
  • For research use only
  • Molecular weight: 285.26
  • Formula: C10H15N5O5
  • Human endogenous metabolite

Catalog No. 50-004-09829


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.