Learn More
Cayman Chemical 2-DeoxygunosIn-5-O-trIpho 5mg
SDP
Supplier:  Cayman Chemical 357715 MG
Encompass_Preferred
A nucleoside triphosphate; has commonly been used in PCR reaction mixtures as a substrate for DNA synthesis by DNA polymerases; can be oxidized to 8-hydroxy-dGTP and 8-oxo-7,8-GTP, which can be misincorporated into DNA
Catalog No. 50-291-1100