Learn More
Apexbio Technology LLC 2-Deoxyinosine 890-38-0 100mg
SDP
Supplier:  Apexbio Technology LLC B7865100
Encompass_Preferred
2-Deoxyinosine (CAS 890-38-0) is a purine nucleoside composed of the base hypoxanthine linked to a deoxyribose sugar Structurally analogous to natural DNA nucleosides 2-deoxyinosine can be incorporated into DNA during replication or repair where it may induce mispairing and promote mutagenesis This property makes it a valuable tool in studies of DNA damage mutational mechanisms and nucleotide metabolism Additionally 2-deoxyinosine is used in enzymatic assays to investigate nucleoside metabolism and the activity of purine nucleoside phosphorylase or related enzymes in biochemical and cellular contexts
Catalog No. 50-000-90913