Learn More
Medchemexpress LLC 2'-deoxyisoguanosine | 106449-56-3 | 99.8% | 267.24 g/mol | C10H13N5O4 | 250 MG

Supplier: Medchemexpress LLC HYW602640250MG
2'-Deoxyisoguanosine is a purine nucleoside analog supplied as a research-grade chemical reagent. It is reported to have low potency and specificity in inhibiting tumor cell growth, similar to other telomerase inhibitors, and is provided with specification and storage guidance for laboratory use.
- High purity (99.8%).
- Molecular weight 267.24 g/mol.
- CAS number 106449-56-3 for unambiguous identification.
- Solid form, light yellow to yellow appearance.
- Recommended storage: powder at -20°C for long-term stability.
- Offered in multiple pack sizes, including 250 MG for preparative experiments.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.