Learn More
Medchemexpress LLC 2-deoxyribose 5-phosphate disodium | 102916-66-5 | MFCD00057568 | 95.0% | C5H9Na2O7P | 10MG
SDP
Supplier:  Medchemexpress LLC HY14706110MG
Encompass_Preferred
2-deoxyribose 5-phosphate disodium is the disodium salt of 2-deoxyribose-5-phosphate, an endogenous metabolite and substrate for 2-deoxyribose-5-phosphate aldolase (DERA). Supplied as a white to off-white solid with 95.0% purity, it is intended for biochemical and enzymatic research applications.
  • Endogenous metabolite and 2-deoxyribose-5-phosphate aldolase (DERA) substrate for enzymatic assays.
  • High purity (95.0%) suitable for research use.
  • Readily soluble in water and DMSO with manufacturer solubility guidance.
  • Stable when stored sealed at -20°C; prepared solutions stable at -80°C for extended storage.
  • Available in small pack sizes appropriate for assay-scale experiments.
Catalog No. 50-002-58191