Learn More
Strem, An Ascensus Company CAS# 1219080-77-9. 1g. 2-(Di-1-adamantylphosphino)dimethylaminobenzene, 97% Me-DalPhos. MFCD17018758
SDP
Supplier:  Strem, An Ascensus Company 1510901G
Encompass
CAS# 1219080-77-9. 1g. 2-(Di-1-adamantylphosphino)dimethylaminobenzene, 97% Me-DalPhos. MFCD17018758. Molecular Weight: 421.60. Molecular Formula: C28H40NP. Color/form: white to yellow xtl. Strem# 15-1090. http://www.strem.com/catalog/v/15-1090/
Catalog No. 50-138-5400