Learn More
Strem, An Ascensus Company CAS# 1160861-60-8. 500mg. 2-Di[3,5-bis(trifluoromethyl)phenylphosphino]-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl, min. 98% JackiePhos. MFCD17015286
SDP
Supplier:  Strem, An Ascensus Company 151157500MG
Encompass
CAS# 1160861-60-8. 500mg. 2-Di[3,5-bis(trifluoromethyl)phenylphosphino]-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl, min. 98% JackiePhos. MFCD17015286. Molecular Weight: 796.66. Molecular Formula: C39H37F12O2P. Color/form: white xtl. Strem# 15-1157.
Catalog No. 50-138-5405