Learn More
Strem, An Ascensus Company CAS# 255837-19-5. 500mg. 2-Di-t-butylphosphino-2'-methyl)-1,1'-biphenyl, 99% t-BuMePhos. MFCD03453047
SDP
Supplier:  Strem, An Ascensus Company 151049500MG
Encompass
CAS# 255837-19-5. 500mg. 2-Di-t-butylphosphino-2'-methyl)-1,1'-biphenyl, 99% t-BuMePhos. MFCD03453047. Molecular Weight: 312.43. Molecular Formula: C21H29P. Color/form: white xtl. Strem# 15-1049. http://www.strem.com/catalog/v/15-1049/
Catalog No. 50-138-5396