Learn More
eMolecules​ 2-Di-tert-butylphosphino-2',4',6'-triisopropylbiphenyl | Combi-Blocks | 564483-19-8 | MFCD06411306 | 424.653 | C29H45P | 98.000 | CC(C)c1cc(C(C)C)c(c(c1)C(C)C)-c1ccccc1P(C(C)(C)C)C(C)(C)C | 5g | 303332261
Supplier:  eMolecules​ 303332261
Encompass_Preferred
2-Di-tert-butylphosphino-2',4',6'-triisopropylbiphenyl | Combi-Blocks | 564483-19-8 | MFCD06411306 | 424.653 | C29H45P | 98.000 | CC(C)c1cc(C(C)C)c(c(c1)C(C)C)-c1ccccc1P(C(C)(C)C)C(C)(C)C | 5g | 303332261
Catalog No. NC3444959