Learn More
Strem, An Ascensus Company CAS# 1049726-96-6. 500mg. 2'-Dicyclohexylphosphino-2,6-dimethoxy-3-sulfonato-1,1'-biphenyl hydrate sodium salt (water soluble SPhos), min. 98%. MFCD07781994
SDP
Supplier:  Strem, An Ascensus Company 151142500MG
Encompass
CAS# 1049726-96-6. 500mg. 2'-Dicyclohexylphosphino-2,6-dimethoxy-3-sulfonato-1,1'-biphenyl hydrate sodium salt (water soluble SPhos), min. 98%. MFCD07781994. Molecular Weight: 512.58. Molecular Formula: C26H34NaO5PS∙XH2O. Color/form: light yellow solid Strem# 15-1142. http://www.strem.com/catalog/v/15-1142/
Catalog No. 50-901-10090