Learn More
Strem, An Ascensus Company CAS# 251320-86-2. 500mg. 2-Dicyclohexylphosphino-2'-methyl-)-1,1'-biphenyl, min. 98% MePhos. MFCD03094577
SDP
Supplier:  Strem, An Ascensus Company 151148500MG
Encompass
CAS# 251320-86-2. 500mg. 2-Dicyclohexylphosphino-2'-methyl-)-1,1'-biphenyl, min. 98% MePhos. MFCD03094577. Molecular Weight: 364.51. Molecular Formula: C25H33P. Color/form: white xtl. Strem# 15-1148. http://www.strem.com/catalog/v/15-1148/
Catalog No. 50-901-10102