Learn More
Strem, An Ascensus Company CAS# 564483-18-7. 2g. 2-(Dicyclohexylphosphino)-2',4',6'-tri-i-propyl-1,1'-biphenyl, min. 98% XPhos. MFCD04117682
SDP
Supplier:  Strem, An Ascensus Company 1511492G
Encompass
CAS# 564483-18-7. 2g. 2-(Dicyclohexylphosphino)-2',4',6'-tri-i-propyl-1,1'-biphenyl, min. 98% XPhos. MFCD04117682. Molecular Weight: 476.72. Molecular Formula: C33H49P. Color/form: white pwdr. Strem# 15-1149. http://www.strem.com/catalog/v/15-1149/
Catalog No. 50-901-10105