Learn More
Apexbio Technology LLC 2-(Dimethylamino)acetaldehyde hydrochloride 125969-54-2 50mg
SDP
Supplier:  Apexbio Technology LLC C700050
Encompass_Preferred
2-Dimethylamino acetaldehyde hydrochloride is a small-molecule intermediate featuring both tertiary amine and aldehyde functional groups It is utilized in the synthesis of cholinergic receptor agonist analogs to facilitate studies of muscarinic acetylcholine receptor activation 2-Dimethylamino acetaldehyde hydrochloride participates in chemical reactions through its electrophilic aldehyde and nucleophilic amine groups enabling the construction of cholinergic mimetic structures relevant to pharmacological research Based on these chemical properties 2-Dimethylamino acetaldehyde hydrochloride holds research potential in exploring receptor-ligand interactions receptor activation profiles and elucidating cholinergic signaling mechanisms
Catalog No. 50-000-91840