Learn More
Medchemexpress LLC 2-(DiMethylphosphoryl)benzenamine | 1197953-47-1 | 169.16 | 50 G

Supplier: Medchemexpress LLC HYW00574650G
2-(DiMethylphosphoryl)benzenamine is a chemical compound that serves as a drug intermediate for the synthesis of various active compounds.
- High purity of 99.85%
- Appearance as a solid
- Color ranges from off-white to light yellow
- Molecular formula C8H12NOP
- Recommended storage at 4°C, protected from light
- For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.