Learn More
Medchemexpress LLC 2-(Diphenylphosphino)pyridine | 37943-90-1 | ≥97.0% | 263.28 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01334350G
Encompass_Preferred
Diphenyl-2-pyridylphosphine is a biochemical reagent that can be utilized as a biological material or organic compound for life science-related research.
  • Can be used as a biological material.
  • Can be used as an organic compound.
  • Applicable for life science related research.
  • Functions as a ligand for metal-catalyzed carbonylations, hydration, dehydrogenative coupling, carbostannylation, distannylation, and silylation.
  • Employed as a reagent for the Mitsunobu reaction.
Catalog No. 50-004-54648