Learn More
Medchemexpress LLC 2-(Diphenylphosphino)pyridine | 37943-90-1 | ≥97.0% | 263.28 | 5 G
Supplier: Medchemexpress LLC HYW0133435G
Diphenyl-2-pyridylphosphine is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is a versatile ligand for various metal-catalyzed reactions, including carbonylations, hydration, dehydrogenative coupling, carbostannylation, distannylation, and silylation. It also serves as a reagent for the Mitsunobu reaction.
- Used as a biochemical reagent
- Can be used for life science-related research
- Functions as a ligand for metal-catalyzed carbonylations
- Also a ligand for hydration and dehydrogenative coupling
- Employed in carbostannylation, distannylation, and silylation reactions
- Reagent for the Mitsunobu reaction