Learn More
Strem, An Ascensus Company CAS# 4848-43-5. 1g. 2-(Diphenylphosphino)ethylamine, min. 95%. MFCD00233831
SDP
Supplier:  Strem, An Ascensus Company 1517621G
Encompass
CAS# 4848-43-5. 1g. 2-(Diphenylphosphino)ethylamine, min. 95%. MFCD00233831. Molecular Weight: 229.26. Molecular Formula: (C6H5)2PCH2CH2NH2. Color/form: colorless to yellow liq. Strem# 15-1762. http://www.strem.com/catalog/v/15-1762/
Catalog No. 50-901-10179