Learn More
Medchemexpress LLC 2-dodecenedioic acid (2E) | 6402-36-4 | 99.8% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11935810MM 1ML
Encompass_Preferred
Traumatic acid (2-dodecenedioic acid) is a plant wound-healing hormone used as a biochemical research reagent. It is supplied as a solid or as a ready-to-use 10 mM solution in DMSO, suitable for studies of wound healing, collagen biosynthesis, apoptosis, and reactive oxygen species.
  • High purity (99.8%) suitable for research applications.
  • Available as solid and as 10 mM solution in DMSO for experimental convenience.
  • Molecular weight 228.28 g/mol and formula C12H20O4 for accurate calculations.
  • Includes cheminformatics identifiers (SMILES) for computational work.
Catalog No. 50-002-33645