Learn More
Cayman Chemical 2-DPMPhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1148110 MG
Encompass_Preferred
Structurally related to pipradrol and methylphenidate (Ritalin), which are psychostimulatory piperadines that inhibit monoamine transporters; identified in recreational drugs; intended for research and forensic applications
Catalog No. 50-295-1494