Learn More
eMolecules​ 617-37-8 | 2-Ethoxy-2-oxoacetic acid | ChemScene | MFCD00785870 | 118.088 | C4H6O4 | 80.000 | CCOC(=O)C(O)=O | 10g | 582640221
Supplier:  eMolecules​ 582640221
Encompass_Preferred
2-Ethoxy-2-oxoacetic acid | ChemScene | 617-37-8 | MFCD00785870 | 118.088 | C4H6O4 | 80.000 | CCOC(=O)C(O)=O | 10g | 582640221
Catalog No. 50-269-5623