Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-ethoxyphenol (guaethol) | 94-71-3 | MFCD00002187 | 99.9% | 138.16 g/mol | C8H10O2 | 500 G
SDP

Supplier:  Medchemexpress LLC HYW022036500G

Encompass_Preferred

2-Ethoxyphenol is a phenolic organic reagent (guaethol) used in biochemical assays and laboratory research. It is a small aromatic ether (C8H10O2) with a molecular weight of 138.16 g/mol, supplied at high reported purity and recommended for controlled laboratory use. Typical handling includes storage at low temperature and protection from air.

  • High reported purity suitable for analytical and research applications.
  • Molecular weight 138.16 g/mol; formula C8H10O2.
  • Appearance: colorless to off-white; solid at lower temperatures.
  • Storage: refrigerate at 4°C under inert atmosphere; in solution store at -80°C for long-term.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-003-01345


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.