Learn More
Medchemexpress LLC 2-Ethyl-6-methylaniline-d13 | 1219794-93-0 | 98.6% | 148.29 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW012626S
Encompass_Preferred
2-Ethyl-6-methylaniline-d13 is the deuterium labeled 2-Ethyl-6-methylaniline. It serves as a tracer and an internal standard for quantitative analysis. The incorporation of stable heavy isotopes into drug molecules, known as deuteration, aids in quantitation during drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules.
  • Aids in quantitation during drug development.
  • Potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-66393