Learn More
Medchemexpress LLC 2-ethylhexyl 2,3,4,5-tetrabromobenzoate | 183658-27-7 | MFCD26960592 | >85.0% | 549.92 g·mol⁻¹ | C15H18Br4O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW33788625MG
Encompass_Preferred
2-ethylhexyl 2,3,4,5-tetrabromobenzoate is a tetrabrominated benzoate used as a flame retardant and supplied in small research-scale quantities. It is a viscous, colorless to light yellow liquid with formula C15H18Br4O2 and molecular weight 549.92 g·mol⁻¹; storage recommendations include -20°C for the pure compound.
  • Used as a flame retardant in materials and analytical research.
  • Available in milligram-scale packs suitable for analytical and small-scale testing.
  • Viscous, colorless to light yellow liquid; soluble in common organic solvents.
  • Stable when stored at -20°C; reduced stability in solution at higher temperatures.
  • Unambiguously identified by CAS number 183658-27-7 for regulatory and safety documentation.
Catalog No. 50-002-81848