Learn More
Sigma Organic Chemistry 2-Ethylhexyl nitrate | 2L | 27247-96-7 | MFCD00011582
Supplier: Sigma Organic Chemistry 2937842.5L
CAS #: 27247-96-7
MDL #: MFCD00011582
Purity: >97 %
UNSPSC Code: 12352100
Molecular Weight: 175.23
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.