Learn More
Sigma Aldrich Fine Chemicals Biosciences 2 Ethylpyrazine 98 FG5KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences W3281035KGK
Encompass
2-Ethylpyrazine is a volatile aroma compound that is mainly formed in food products such as roasted coffee beans or roasted sesame seeds due to the Maillard reaction between sugars and proteins during the roasting process.
Catalog No. 50-290-8347