Learn More
Apexbio Technology LLC 2'-F-dCTP(Synonyms: 2'-Fluoro-2'-deoxycytidine-5'-triphosphate, 2'-Fluoro-dCTP, 2'-F-2'-deoxycytidine triphosphate), 10ul (100 mM).
SDP
Supplier:  Apexbio Technology LLC B79610.01
Encompass_Preferred
2 -F-dCTP is a small molecule analog of deoxycytidine triphosphate that exerts its biological effects primarily through modulation of nucleotide metabolic pathways It is known to interfere with specific molecular signaling cascades and enzymatic activities involved in DNA synthesis and metabolism Due to its ability to stably modulate the activities of relevant biological targets 2 -F-dCTP has been utilized in biomedical research to investigate and disrupt metabolic pathways linked to disease mechanisms aiding in the identification of novel therapeutic approaches
Catalog No. 50-000-61235