Learn More
Apexbio Technology LLC 2'-F-dGTP(Synonyms: 2'-Fluoro-2'-deoxyguanosine-5'-triphosphate, 2'-Fluoro-dGTP, 2'-F-2'-deoxyguanosine triphosphate), 5x100ul (100 mM).
SDP
Supplier:  Apexbio Technology LLC B79620.5
Encompass_Preferred
2 -F-dGTP (2 -Fluoro-2 -deoxyguanosine triphosphate) is a chemically modified deoxynucleotide analog commonly employed in nucleic acid research It contains a fluorine substitution at the 2 position of the deoxyribose sugar moiety enabling its incorporation into DNA by polymerases The incorporation of this modified nucleotide confers enhanced resistance to nuclease-mediated degradation improving nucleic acid stability Researchers frequently use 2 -F-dGTP to investigate polymerase specificity enzymatic mechanism and to develop antisense oligonucleotides and aptamers with increased biological durability
Catalog No. 50-000-60879