Learn More
Medchemexpress LLC 2-Fluoro-5-formylbenzonitrile | 218301-22-5 | 99.7% | 149.12 | 25 G

Supplier: Medchemexpress LLC 25GHY20138
2-Fluoro-5-formylbenzonitrile is a drug intermediate for synthesis of various active compounds. It is for research use only.
- Purity of 99.67%
- Molecular weight of 149.12
- Formula C8H4FNO
- Appearance as a solid
- Color is off-white to light yellow
- Ships at room temperature in continental US
- Store at 4°C, under nitrogen
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.