Learn More
Cayman Chemical 2-FluoroadenIn 250mg
SDP
Supplier:  Cayman Chemical 30421250 MG
Encompass_Preferred
A heterocyclic building block; has been used in the synthesis of Hsp90 inhibitors that have anticancer activity in vitro; cytotoxic to CEM human leukemia cells (IC50 = 0.15 µM)
Catalog No. 50-299-1828