Learn More
Medchemexpress LLC 2-fluoropalmitic acid | 16518-94-8 | 98.0% | 274.41 g·mol⁻¹ | C16H31FO2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1176515MG
Encompass_Preferred
2-Fluoropalmitic acid is a fluorinated long-chain fatty acid used as a research reagent to probe fatty-acid metabolism and acyl-CoA processing. It acts as an acyl-CoA synthetase inhibitor and is employed in biochemical studies investigating lipid metabolism and enzyme activity.
  • Inhibits acyl-CoA synthetase activity for metabolic and enzymatic assays.
  • High reported purity suitable for research applications.
  • Small quantity packaging appropriate for assay development and pilot studies.
  • Fluorinated analog of palmitic acid for use in metabolic pathway probing.
Catalog No. 50-002-28119