Learn More
Medchemexpress LLC 2-fluoropalmitic acid | 16518-94-8 | MFCD00216145 | 98.0% | 274.41 g/mol | C16H31FO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11765110MG
Encompass_Preferred
2-Fluoropalmitic acid is a fluorinated long-chain fatty acid that functions as an acyl-CoA synthetase inhibitor and has been reported as a candidate anti-glioma agent that suppresses viability, stem-like phenotype, proliferation, and invasion of glioma cell lines. Supplied for research use with a reported purity of 98.0%.
  • Acts as an acyl-CoA synthetase inhibitor
  • Reported to suppress viability and stem-like phenotype of glioma stem cells
  • Reported to inhibit proliferation and invasion of glioma cell lines
  • High purity (98.0%) suitable for research use
  • Available as a 10 mg quantity for biochemical and cell-based studies
  • Molecular weight 274.41 g/mol; chemical formula C16H31FO2
Catalog No. 50-002-28788